Learn More
(2R)-2-(6-Methoxynaphthalen-2-yl)propanoic Acid ((R)-Naproxen), Mikromol™
Description
(2R)-2-(6-Methoxynaphthalen-2-yl)propanoic Acid ((R)-Naproxen)
Specifications
Specifications
| Chemical Name or Material | (R)-Naproxen |
| CAS | 23979-41-1 |
| Analyte or Component Names | (R)-Naproxen |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H14 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Naproxen Imp. G (EP), Naproxen USP Related Compound G, Naproxen USP RC G, (2R)-2-(6-Methoxynaphthalen-2-yl)propanoic Acid, R-Naproxen, (alphaR)-6-methoxy-alpha-methyl-2-naphthaleneacetic acid, 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (R)- (8CI), (-)-(R)-2-(6-Methoxy-2-naphthyl)propionic acid, (-)-2-(6-Methoxy-2-naphthyl)propionic acid, (-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid, (-)-Naproxen, (R)-(-)-Naproxen, (R)-2-(6-Methoxy-2-naphthyl)propionic acid, (R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen, (alphaR)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid, l-Naproxen, Naproxen Related Compound G, Naproxen Impurity G |
| InChI Formula | InChI=1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m1/s1 |
| SMILES | COc1ccc2cc(ccc2c1)[C@@H](C)C(=O)O |
| IUPAC Name | (2R)-2-(6-methoxynaphthalen-2-yl)propanoic acid |
| Show More |