missing translation for 'onlineSavingsMsg'
Learn More

Pemetrexed Dimethyl Ester, Mikromol™

Product Code. 30051481
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30051481

Brand: Mikromol MM1301.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pemetrexed Dimethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pemetrexed Dimethyl Ester
CAS 155405-81-5
Analyte or Component Names Pemetrexed Dimethyl Ester
Shipping Condition Room Temperature
Molecular Formula C22 H25 N5 O6
Certifications/Compliance ISO 9001
Synonym L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-, 1,5-dimethyl ester, L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-, dimethyl ester (9CI), 1H-Pyrrolo[2,3-d]pyrimidine, L-glutamic acid deriv.
InChI Formula InChI=1S/C22H25N5O6/c1-32-16(28)10-9-15(21(31)33-2)25-19(29)13-6-3-12(4-7-13)5-8-14-11-24-18-17(14)20(30)27-22(23)26-18/h3-4,6-7,11,15H,5,8-10H2,1-2H3,(H,25,29)(H4,23,24,26,27,30)/t15-/m0/s1
SMILES COC(=O)CC[C@H](NC(=O)c1ccc(CCc2c[nH]c3NC(=NC(=O)c23)N)cc1)C(=O)OC
IUPAC Name dimethyl (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioate
Molecular Weight (g/mol) 455.4638
Quantity 25 mg
Formula Weight 455.1805 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.