Learn More
Acebutolol EP Impurity I, Mikromol™
Description
Acebutolol EP Impurity I
Specifications
Specifications
| Chemical Name or Material | N-[3-Acetyl-4-[(2RS)-3-(ethylamino)-2-hydroxypropoxy]phenyl]butanamide |
| CAS | 441019-91-6 |
| Analyte or Component Names | N-[3-Acetyl-4-[(2RS)-3-(ethylamino)-2-hydroxypropoxy]phenyl]butanamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H26 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Acebutolol Hydrochloride Imp. I (EP), Acebutolol Imp. I (EP), Acebutolol USP Related Compound I, Acebutolol USP RC I, N-[3-Acetyl-4-[(2RS)-3-(ethylamino)-2-hydroxypropoxy]phenyl]butanamide, Acebutolol Related Compound I, Acebutolol Hydrochloride Impurity I, Acebutolol Impurity I |
| InChI Formula | InChI=1S/C17H26N2O4/c1-4-6-17(22)19-13-7-8-16(15(9-13)12(3)20)23-11-14(21)10-18-5-2/h7-9,14,18,21H,4-6,10-11H2,1-3H3,(H,19,22) |
| SMILES | CCCC(=O)Nc1ccc(OCC(O)CNCC)c(c1)C(=O)C |
| IUPAC Name | N-[3-acetyl-4-[3-(ethylamino)-2-hydroxypropoxy]phenyl]butanamide |
| Show More |