Learn More
Mefenamic Acid EP Impurity B, Mikromol™
Description
Mefenamic Acid EP Impurity B
Specifications
Specifications
| Chemical Name or Material | N-(2,3-Dimethylphenyl)-2-[(2,3-dimethylphenyl)amino]benzamide |
| CAS | 21122-68-9 |
| Analyte or Component Names | N-(2,3-Dimethylphenyl)-2-[(2,3-dimethylphenyl)amino]benzamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H24 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Mefenamic Acid Imp. B (EP), N-(2,3-Dimethylphenyl)-2-[(2,3-dimethylphenyl)amino]benzamide, Mefenamic Acid Impurity B |
| InChI Formula | InChI=1S/C23H24N2O/c1-15-9-7-13-20(17(15)3)24-22-12-6-5-11-19(22)23(26)25-21-14-8-10-16(2)18(21)4/h5-14,24H,1-4H3,(H,25,26) |
| SMILES | Cc1cccc(NC(=O)c2ccccc2Nc3cccc(C)c3C)c1C |
| IUPAC Name | 2-(2,3-dimethylanilino)-N-(2,3-dimethylphenyl)benzamide |
| Show More |