missing translation for 'onlineSavingsMsg'
Learn More

HS-173, MedChemExpress

Product Code. 16433506
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433506 25 mg 25mg
16486878 10 mg 10mg
16469417 5 mg 5mg
16492937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16433506 Supplier Medchem Express Supplier No. HY15868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HS-173 is a novel PI3K inhibitor, that is used for cancer treatment.

HS-173 is a novel PI3K inhibitor, that is used for cancer treatment.

Specifications

Chemical Name or Material HS-173
CAS 1276110-06-5
Color Gray
Purity Grade Notes Research
Molecular Formula C21H18N4O4S
Quantity 25 mg
Solubility Information DMSO : 50 mg/mL (118.35 mM; Need ultrasonic)
SMILES O=C(C1=CN=C2C=CC(C3=CC(NS(=O)(C4=CC=CC=C4)=O)=CN=C3)=CN21)OCC
Molecular Weight (g/mol) 422.46
Formula Weight 422.46
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.