missing translation for 'onlineSavingsMsg'
Learn More

(4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines), Mikromol™

Product Code. 30050275
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050275 25 mg 25mg
17208509 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050275 Supplier Mikromol Supplier No. MM0232.030025
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines)
CAS 94659-47-9
Analyte or Component Names (4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines)
Shipping Condition Room Temperature
Molecular Formula C16 H19 N3 O5 S
Certifications/Compliance ISO 17034
Synonym Amoxicillin Trihydrate Imp. C (EP), Amoxicillin Imp. C (EP), Amoxicillin USP Related Compound C, Amoxicillin USP RC C, (4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid, Amoxicillin Diketopiperazines, SB-206891, Ph Eur Amoxicillin Impurity C, Amoxicillin Related Compound C, Amoxicillin Trihydrate Impurity C, Amoxicillin Impurity C
InChI Formula InChI=1S/C16H19N3O5S/c1-16(2)11(15(23)24)19-14(25-16)10-13(22)17-9(12(21)18-10)7-3-5-8(20)6-4-7/h3-6,9-11,14,19-20H,1-2H3,(H,17,22)(H,18,21)(H,23,24)/t9?,10?,11-,14?/m0/s1
SMILES CC1(C)SC(N[C@H]1C(=O)O)C2NC(=O)C(NC2=O)c3ccc(O)cc3
IUPAC Name (4S)-2-[5-(4-hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol) 365.4
Quantity 25 mg
Formula Weight 365.1045 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.