missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine (Depramine; 10,11-Dehydroimipramine), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine (Depramine; 10,11-Dehydroimipramine)
Specifications
Specifications
| Chemical Name or Material | 3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine (Depramine; 10,11-Dehydroimipramine) |
| CAS | 303-54-8 |
| Analyte or Component Names | 3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine (Depramine; 10,11-Dehydroimipramine) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 N2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Imipramine Hydrochloride Imp. B (EP), Imipramine Imp. B (EP), Depramine, 3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine, 10,11-Dehydroimipramine, 5H-Dibenz[b,f]azepine-5-propanamine, N,N-dimethyl-, 5H-Dibenz[b,f]azepine, 5-[3-(dimethylamino)propyl]- (6CI,7CI,8CI), N,N-Dimethyl-5H-dibenz[b,f]azepine-5-propanamine, 5-[3-(Dimethylamino)propyl]-5H-dibenz[b,f]azepine, Balipramine, G 31406, GP 31406, Imipramine Hydrochloride Impurity B, Imipramine Impurity B |
| InChI Formula | InChI=1S/C19H22N2/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21/h3-6,8-13H,7,14-15H2,1-2H3 |
| SMILES | CN(C)CCCN1c2ccccc2C=Cc3ccccc13 |
| IUPAC Name | 3-benzo[b][1]benzazepin-11-yl-N,N-dimethylpropan-1-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction