missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-2-(Benzylmethylamino)-1-(3-hydroxyphenyl)ethanol (Benzylphenylephrine), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(1R)-2-(Benzylmethylamino)-1-(3-hydroxyphenyl)ethanol (Benzylphenylephrine)
Specifications
Specifications
| Chemical Name or Material | (1R)-2-(Benzylmethylamino)-1-(3-hydroxyphenyl)ethanol |
| CAS | 1367567-95-0 |
| Analyte or Component Names | (1R)-2-(Benzylmethylamino)-1-(3-hydroxyphenyl)ethanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1R)-2-(Benzylmethylamino)-1-(3-hydroxyphenyl)ethanol, Benzylphenylephrine, (R)-3-{2-[Benzyl(methyl)amino]-1-hydroxyethyl}phenol, Phenylephrine Related Compound D, Phenylephrine Hydrochloride Impurity D, Phenylephrine Impurity D, Phenylephrine Impurity D |
| InChI Formula | InChI=1S/C16H19NO2/c1-17(11-13-6-3-2-4-7-13)12-16(19)14-8-5-9-15(18)10-14/h2-10,16,18-19H,11-12H2,1H3/t16-/m0/s1 |
| SMILES | CN(C[C@H](O)c1cccc(O)c1)Cc2ccccc2 |
| IUPAC Name | 3-[(1R)-2-[benzyl(methyl)amino]-1-hydroxyethyl]phenol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction