missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Standard: Mg @ 1000 μg/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
Description
Magnesium Standard: Mg @ 1000 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Magnesium |
| CAS | 7439-95-4 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Magnesium |
| Shipping Condition | Room Temperature |
| Molecular Formula | Mg |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Mg |
| InChI Formula | InChI=1S/Mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction