missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclizine Hydrochloride, Mikromol™
Description
Cyclizine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Cyclizine Hydrochloride |
| CAS | 303-25-3 |
| Analyte or Component Names | Cyclizine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Cyclizine hydrochloride, 1-(Diphenylmethyl)-4-methylpiperazine hydrochloride, Piperazine, 1-(diphenylmethyl)-4-methyl-, hydrochloride (1:1), Piperazine, 1-(diphenylmethyl)-4-methyl-, monohydrochloride (8CI,9CI), Marezine hydrochloride, Marzine, N-Benzhydryl-N'-methylpiperazine monohydrochloride |
| InChI Formula | InChI=1S/C18H22N2.ClH/c1-19-12-14-20(15-13-19)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17;/h2-11,18H,12-15H2,1H3;1H |
| SMILES | Cl.CN1CCN(CC1)C(c2ccccc2)c3ccccc3 |
| IUPAC Name | 1-benzhydryl-4-methylpiperazine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?