missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefuroxime Sodium, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Cefuroxime Sodium
Specifications
Specifications
| Chemical Name or Material | Cefuroxime Sodium |
| CAS | 56238-63-2 |
| Analyte or Component Names | Cefuroxime Sodium |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H15 N4 O8 S . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Cefuroxime Axetil Imp. D (EP), Cefuroxime sodium, Sodium (6R,7R)-3-[(carbamoyloxy)methyl]-7-[[(Z)-(furan-2-yl)(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, 750/2, 640/3, 640/68, 640/1, 640/2, GSK682709A, SD, SC, SB, 750/359, SA, Cefuroxime Sodium (640/359), GSK682708A |
| InChI Formula | InChI=1S/C16H16N4O8S.Na/c1-26-19-9(8-3-2-4-27-8)12(21)18-10-13(22)20-11(15(23)24)7(5-28-16(17)25)6-29-14(10)20;/h2-4,10,14H,5-6H2,1H3,(H2,17,25)(H,18,21)(H,23,24);/q;+1/p-1/b19-9-;/t10-,14-;/m1./s1 |
| SMILES | [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])COC(=O)N)\c3occc3 |
| IUPAC Name | sodium;(6R,7R)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction