missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Dobesilate Monohydrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Calcium Dobesilate Monohydrate
Specifications
Specifications
| Chemical Name or Material | Calcium Dobesilate Monohydrate |
| Analyte or Component Names | Calcium Dobesilate Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C6 H5 O5 S . Ca . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Calcium Dobesilate Monohydrate, Calcium di(2,5-dihydroxybenzenesulfonate) monohydrate |
| InChI Formula | InChI=1S/2C6H6O5S.Ca.H2O/c2*7-4-1-2-5(8)6(3-4)12(9,10)11;;/h2*1-3,7-8H,(H,9,10,11);;1H2/q;;+2;/p-2 |
| SMILES | O.[Ca+2].Oc1ccc(O)c(c1)S(=O)(=O)[O-].Oc2ccc(O)c(c2)S(=O)(=O)[O-] |
| IUPAC Name | calcium;2,5-dihydroxybenzenesulfonate;hydrate |
| Molecular Weight (g/mol) | 436.43 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction