Learn More
25-Hydroxy VD2-d6, MedChemExpress
Product Code. 16439047
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439047 5 mg 5mg
16401509 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16439047 Supplier Medchem Express Supplier No. HY15328
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress 25-Hydroxy VD2-d6 is the deuterium labeled 25-Hydroxy VD2.
25-Hydroxy VD2-d6 is the deuterium labeled 25-Hydroxy VD2.
Specifications
Chemical Name or Material 25-Hydroxy VD2-d6
CAS 1262843-46-8
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C28H38D6O2
Color White
Solubility Information DMSO : 100 mg/mL (238.85 mM; Need ultrasonic)
SMILES O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2
Molecular Weight (g/mol) 418.68
Quantity 5 mg
Formula Weight 418.68
Percent Purity 98.96%
Grade Research