missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzyl chloride, 98%, Thermo Scientific™

Product Code. p-7030753
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454197 1 g 1g
15464197 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454197 Supplier Thermo Scientific Alfa Aesar Supplier No. H31603.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116827-40-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD00221456
InChI Key KSHPWYXAJJCUMJ-UHFFFAOYSA-N
Synonym 1-chloromethyl-2-trifluoromethoxy benzene, 2-trifluoromethoxy benzyl chloride, benzene,1-chloromethyl-2-trifluoromethoxy, acmc-20dw7r, 2-trifluoromethoxybenzyl chloride, 2-trifluoromethyoxy benzyl chloride, 2-chloromethyl phenoxy trifluoromethane
PubChem CID 4656677
IUPAC Name 1-(chloromethyl)-2-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC=C1C(Cl)=O

Specifications

Quantity 1 g
UN Number UN3265
Formula Weight 210.58
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethoxy)benzyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.