accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFOClO

CAS RN 116827-40-8

IUPAC Name: 2-(trifluoromethoxy)benzoyl chloride
Synonyms: 2-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: KSHPWYXAJJCUMJ-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results