missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl propionate, 97%, Thermo Scientific Chemicals

Product Code. 11469187
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469187 250 g 250g
11459187 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469187 Supplier Thermo Scientific Alfa Aesar Supplier No. B21283.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-70-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00027008
InChI Key HVGZQCSMLUDISR-UHFFFAOYSA-N
Synonym 2-phenylethyl propionate, phenethyl propionate, propanoic acid, 2-phenylethyl ester, phenylethyl propionate, 2-phenethyl propionate, benzylcarbinyl propionate, ecopco acu, phenethyl alcohol, propionate, phenethyl popanoate, japanese beetle bait
PubChem CID 31225
IUPAC Name 2-phenylethyl propanoate
SMILES CCC(=O)OCCC1=CC=CC=C1

Specifications

Density 1.007
Boiling Point 245°C
Flash Point 106°C (223°F)
Odor Floral
Refractive Index 1.494
Quantity 250 g
Formula Weight 178.22
Percent Purity 97%
Chemical Name or Material 2-Phenylethyl propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.