accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COO

CAS RN 122-70-3

IUPAC Name: 2-phenylethyl propanoate
Synonyms: 2-phenylethyl propionate propionic acid, phenethyl ester mixed with eugen
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: HVGZQCSMLUDISR-UHFFFAOYSA-N
SMILES: CCC(=O)OCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results