Learn More
2-Pentynoic acid, 97%
Product Code. p-7035687
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 15490188
Brand: Thermo Scientific Alfa Aesar H53411.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 5963-77-9 | C5H6O2 | 98.101 g/mol
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5963-77-9
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00014370
InChI Key MINRDQDGBLQBGD-UHFFFAOYSA-N
Synonym 2-pentynoic acid, pentynoic acid, 2-pentynoicacid, 2-pentyneoic acid, acmc-1ay8s
PubChem CID 324379
IUPAC Name pent-2-ynoic acid
SMILES CCC#CC(=O)O
Specifications
Melting Point 47°C to 53°C
Flash Point 96°C (205°F)
Linear Formula CH3CH2CH≡CCO2H
Quantity 1 g
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material 2-Pentynoic acid