accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5963-77-9

CH3HOO

CAS RN 5963-77-9

IUPAC Name: pent-2-ynoic acid
Synonyms: pent-2-ynoic acid
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: MINRDQDGBLQBGD-UHFFFAOYSA-N
SMILES: CCC#CC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results