missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)benzyl alcohol, 97+%, Thermo Scientific™

Product Code. 15494147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494147 5 g 5g
15484147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494147 Supplier Thermo Scientific Alfa Aesar Supplier No. H31559.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as important intermediates and reagents in organic synthesis. Also used as solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133605-27-3
Molecular Formula C8H6F3NO3
Molecular Weight (g/mol) 221.14
MDL Number MFCD03095418
InChI Key FQPWWEYTNHARPU-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl phenyl methanol, 2-nitro-4-trifluoromethyl benzyl alcohol, 2-nitro-4-trifluoromethylphenyl methanol
PubChem CID 45790938
IUPAC Name [2-nitro-4-(trifluoromethyl)phenyl]methanol
SMILES OCC1=C(C=C(C=C1)C(F)(F)F)[N+]([O-])=O

Specifications

Melting Point 63°C to 65°C
Quantity 5 g
Formula Weight 221.13
Percent Purity ≥97%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.