accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFNOOOH

CAS RN 133605-27-3

IUPAC Name: [2-nitro-4-(trifluoromethyl)phenyl]methanol
Synonyms: [2-nitro-4-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 221.14
Molecular Formula: C8H6F3NO3
InChi Key: FQPWWEYTNHARPU-UHFFFAOYSA-N
SMILES: OCC1=C(C=C(C=C1)C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results