missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. p-7032837
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15468955 250 mg 250mg
15458955 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15468955

Brand: Thermo Scientific Alfa Aesar H34468.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use as chiral derivatizing agents in the determination of the enantiomeric excess of alcohols and amines. They are widely used as important intermediates in the synthesis of dyes, drugs and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as chiral derivatizing agents in the determination of the enantiomeric excess of alcohols and amines. They are widely used as important intermediates in the synthesis of dyes, drugs and pigments.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Water, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257535-05-9
Molecular Formula C7H5ClF3NO2S
Molecular Weight (g/mol) 259.63
MDL Number MFCD14583150
InChI Key BNCNLCKWRCXBQH-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-2-methyl-6-trifluoromethyl pyridine
PubChem CID 51342031
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
SMILES CC1=C(C=CC(=N1)C(F)(F)F)S(Cl)(=O)=O

Specifications

Melting Point 57°C to 59°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 259.64
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.