accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1257535-05-9

IUPAC Name: 2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Synonyms: 2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Molecular Weight (g/mol): 259.63
Molecular Formula: C7H5ClF3NO2S
InChi Key: BNCNLCKWRCXBQH-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(=N1)C(F)(F)F)S(Cl)(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results