Learn More
2-Methyl-3-vinyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride, Mikromol
Product Code. 30462986
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462986 4 x 25 mg 100mg
30052270 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30462986 Supplier Mikromol Supplier No. MM0491.3400254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Methyl-3-vinyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methyl-3-vinyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride
CAS 2726145-01-1
Analyte or Component Names 2-Methyl-3-vinyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C11 H14 N2 O . Cl H
Certifications/Compliance ISO 17025
Synonym 2-Methyl-3-vinyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one Hydrochloride
InChI Formula InChI=1S/C11H14N2O.ClH/c1-3-9-8(2)12-10-6-4-5-7-13(10)11(9)14;/h3H,1,4-7H2,2H3;1H
Solution Type Neat
SMILES Cl.CC1=C(C=C)C(=O)N2CCCCC2=N1
IUPAC Name 3-ethenyl-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one;hydrochloride
Molecular Weight (g/mol) 226.7
Quantity 4 x 25 mg
Formula Weight 226.0873 g/mol