Learn More
2-Methyl-1,8-naphthyridine, 97%, Thermo Scientific Chemicals
Product Code. 15416937
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416937 1 g 1g
15426937 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15416937 Supplier Thermo Scientific Alfa Aesar Supplier No. H34032.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 1569-16-0
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD04066717
InChI Key FSWRUYCICUXURT-UHFFFAOYSA-N
Synonym 2-methyl-1,8 naphthyridine, 2-methyl 1,8-naphthyridine, 2-methyl-1,8-naphthyridine, 1,8-naphthyridine, 2-methyl, 2-methylpyridino 2,3-b pyridine, 2-methyl-1,8-naphtyridine, 2-methyl 1,8 naphthyridine, methylnaphthyridine, 2-methylnaphthyridine
PubChem CID 74073
IUPAC Name 2-methyl-1,8-naphthyridine
SMILES CC1=NC2=C(C=CC=N2)C=C1
Specifica
Melting Point 91°C to 93°C
Quantity 1 g
Formula Weight 144.18
Percent Purity 97%
Chemical Name or Material 2-Methyl-1,8-naphthyridine
compliance-icons
Identificativo prodotto
  • 2-Methyl-1,8-naphthyridine
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Informazioni supplementari
  • MIXTURE LIST-Contain: 2-Methyl-1,8-naphthyridine
RUO – Research Use Only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.