accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNN

CAS RN 1569-16-0

IUPAC Name: 2-methyl-1,8-naphthyridine
Synonyms: 2-methyl-1,8-naphthyridine
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: FSWRUYCICUXURT-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=NC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results