missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,8-naphthyridine, 97%, Thermo Scientific Chemicals

Product Code. 15426937
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426937 250 mg 250mg
15416937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426937 Supplier Thermo Scientific Alfa Aesar Supplier No. H34032.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1569-16-0
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD04066717
InChI Key FSWRUYCICUXURT-UHFFFAOYSA-N
Synonym 2-methyl-1,8 naphthyridine, 2-methyl 1,8-naphthyridine, 2-methyl-1,8-naphthyridine, 1,8-naphthyridine, 2-methyl, 2-methylpyridino 2,3-b pyridine, 2-methyl-1,8-naphtyridine, 2-methyl 1,8 naphthyridine, methylnaphthyridine, 2-methylnaphthyridine
PubChem CID 74073
IUPAC Name 2-methyl-1,8-naphthyridine
SMILES CC1=NC2=C(C=CC=N2)C=C1

Specifications

Melting Point 91°C to 93°C
Quantity 250 mg
Formula Weight 144.18
Percent Purity 97%
Chemical Name or Material 2-Methyl-1,8-naphthyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.