missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,4-bis(trimethylsiloxy)benzene, 98%, Thermo Scientific™

Product Code. p-7026226
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11499627

Brand: Thermo Scientific Alfa Aesar B22116.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78018-53-8
Molecular Formula C13H24O2Si2
Molecular Weight (g/mol) 268.50
MDL Number MFCD02683083
InChI Key IPYOHMJSEPFQSQ-UHFFFAOYSA-N
Synonym 2-methyl-1,4-bis trimethylsiloxy benzene, trimethyl 3-methyl-4-trimethylsilyl oxy phenoxy silane, 2,5-bis trimethylsiloxy toluene, methylhydroquinone, 2tms derivative, methylhydroquinone, bis trimethylsilyl ether, 1,4-bis trimethylsilyl oxy-2-methylbenzene, benzene,2-methyl-1,4-bis trimethylsilyl oxy, 2-methyl-1,4-phenylene bis oxy bis trimethylsilane
PubChem CID 12691803
IUPAC Name trimethyl-(2-methyl-4-trimethylsilyloxyphenoxy)silane
SMILES CC1=C(O[Si](C)(C)C)C=CC(O[Si](C)(C)C)=C1

Specifications

Quantity 5 g
Formula Weight 268.51
Percent Purity 98%
Sensitivity Moisture sensitive
Chemical Name or Material 2-Methyl-1,4-bis(trimethylsiloxy)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.