accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 78018-53-8

CH3OSiH3CH3CCH3OSiCH3CH3H3C

CAS RN 78018-53-8

IUPAC Name: trimethyl({2-methyl-4-[(trimethylsilyl)oxy]phenoxy})silane
Synonyms: trimethyl(2-methyl-4-[(trimethylsilyl)oxy]phenoxy)silane
Molecular Weight (g/mol): 268.5
Molecular Formula: C13H24O2Si2
InChi Key: IPYOHMJSEPFQSQ-UHFFFAOYSA-N
SMILES: CC1=C(O[Si](C)(C)C)C=CC(O[Si](C)(C)C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results