Learn More
2-Isopropoxypyridine-3-carboxaldehyde, 98%
Product Code. 11474510
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11474510
Brand: Thermo Scientific Alfa Aesar H50089.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 885278-10-4 | C9H11NO2 | 165.19 g/mol
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaceutical intermediate.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 885278-10-4
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD06804560
InChI Key GNTQOKGIVMJHQG-UHFFFAOYSA-N
Synonym 2-isopropoxynicotinaldehyde, 2-isopropoxypyridine-3-carboxaldehyde, 2-isopropoxy-pyridine-3-carbaldehyde, 2-isopropoxypyridine-3-carbaldehyde, 2-propan-2-yl-oxy pyridine-3-carbaldehyde, acmc-20anso, 2-methylethoxy pyridine-3-carbaldehyde, 2-propan-2-yloxy pyridine-3-carbaldehyde, 2-propan-2-yl oxy pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde,2-1-methylethoxy
PubChem CID 42553128
IUPAC Name 2-propan-2-yloxypyridine-3-carbaldehyde
SMILES CC(C)OC1=C(C=O)C=CC=N1
Specifications
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 2-Isopropoxypyridine-3-carboxaldehyde