CAS RN 885278-10-4
CAS RN 885278-10-4
IUPAC Name:
2-(propan-2-yloxy)pyridine-3-carbaldehyde
Synonyms:
2-isopropoxypyridine-3-carbaldehyde
Molecular Weight (g/mol):
165.19
Molecular Formula:
C9H11NO2
InChi Key:
GNTQOKGIVMJHQG-UHFFFAOYSA-N
SMILES:
CC(C)OC1=C(C=O)C=CC=N1
1
–
1
of
1
results
1
2-Isopropoxypyridine-3-carboxaldehyde, 98%
CAS: 885278-10-4 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 MDL Number: MFCD06804560 InChI Key: GNTQOKGIVMJHQG-UHFFFAOYSA-N Synonym: 2-isopropoxynicotinaldehyde,2-isopropoxypyridine-3-carboxaldehyde,2-isopropoxy-pyridine-3-carbaldehyde,2-isopropoxypyridine-3-carbaldehyde,2-propan-2-yl-oxy pyridine-3-carbaldehyde,acmc-20anso,2-methylethoxy pyridine-3-carbaldehyde,2-propan-2-yloxy pyridine-3-carbaldehyde,2-propan-2-yl oxy pyridine-3-carbaldehyde,3-pyridinecarboxaldehyde,2-1-methylethoxy PubChem CID: 42553128 IUPAC Name: 2-propan-2-yloxypyridine-3-carbaldehyde SMILES: CC(C)OC1=C(C=O)C=CC=N1