Learn More
2-(Furan-2-yl)-2-oxoacetic Acid, Mikromol™
Description
2-(Furan-2-yl)-2-oxoacetic Acid
Specifications
Specifications
| Chemical Name or Material | alpha-Oxo-2-furanacetic Acid |
| CAS | 1467-70-5 |
| Analyte or Component Names | alpha-Oxo-2-furanacetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6H4O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-(2-Furyl)-2-oxoacetic acid, 2-(Furan-2-yl)-2-oxoacetic acid, 2-Furanyl(oxo)acetic acid, 2-Furylglyoxylic acid, 2-Oxo-2-(2-furanyl)acetic acid, Furoylformic acid, NSC 59069, 2-Furanglyoxylic acid, 2-Furanacetic acid, α-oxo-, 2-Furanglyoxylic acid (7CI,8CI), α-Oxo-2-furanacetic acid |
| InChI Formula | InChI=1S/C6H4O4/c7-5(6(8)9)4-2-1-3-10-4/h1-3H,(H,8,9) |
| SMILES | OC(=O)C(=O)c1occc1 |
| IUPAC Name | 2-(furan-2-yl)-2-oxoacetic acid |
| Show More |