Learn More
2-Ethylhexyl 2-(3-Benzoylphenyl)propionate (Ketoprofen 2-Ethylhexyl Ester), Mikromol™
Description
2-Ethylhexyl 2-(3-Benzoylphenyl)propionate (Ketoprofen 2-Ethylhexyl Ester)
Specifications
Specifications
| Chemical Name or Material | Ketoprofen 2-Ethylhexyl Ester |
| CAS | 2731997-93-4 |
| Analyte or Component Names | Ketoprofen 2-Ethylhexyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H30 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Ethylhexyl 2-(3-Benzoylphenyl)propionate, Ketoprofen 2-Ethylhexyl Ester, 2-Ethylhexyl 3-benzoyl-α-methylbenzeneacetate (ACI), Benzeneacetic acid, 3-benzoyl-α-methyl-, 2-ethylhexyl ester (ACI) |
| InChI Formula | InChI=1S/C24H30O3/c1-4-6-11-19(5-2)17-27-24(26)18(3)21-14-10-15-22(16-21)23(25)20-12-8-7-9-13-20/h7-10,12-16,18-19H,4-6,11,17H2,1-3H3 |
| SMILES | CCCCC(CC)COC(=O)C(C)c1cccc(c1)C(=O)c2ccccc2 |
| IUPAC Name | 2-ethylhexyl 2-(3-benzoylphenyl)propanoate |
| Show More |