Learn More
2-Ethyl-2-phenalacetylguanidine Sulphate, Mikromol™
Product Code. 30460075
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460075 4 x 25 mg -
17902986 100 mg 100mg
17983186 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30460075 Supplier Mikromol Supplier No. MM0265.1000254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Ethyl-2-phenalacetylguanidine Sulphate, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Ethyl-2-phenalacetylguanidine Sulphate
CAS 2732673-27-5
Analyte or Component Names 2-Ethyl-2-phenalacetylguanidine Sulphate
Shipping Condition Room Temperature
Molecular Formula 2 C11 H15 N3 O . H2 O4 S
Certifications/Compliance ISO 17025
Synonym 2-Ethyl-2-phenalacetylguanidine Sulphate
InChI Formula InChI=1S/2C11H15N3O.H2O4S/c2*1-2-9(10(15)14-11(12)13)8-6-4-3-5-7-8;1-5(2,3)4/h2*3-7,9H,2H2,1H3,(H4,12,13,14,15);(H2,1,2,3,4)
Solution Type Neat
SMILES CCC(C(=O)NC(=N)N)c1ccccc1.CCC(C(=O)NC(=N)N)c2ccccc2.OS(=O)(=O)O
IUPAC Name N-carbamimidoyl-2-phenylbutanamide;sulfuric acid
Molecular Weight (g/mol) 508.59
Quantity 4 x 25 mg
Formula Weight 508.2104 g/mol