missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-4-(tri-n-butylstannyl)thiazole, 96%, Thermo Scientific™

Product Code. p-7034003
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488107 250 mg 250mg
15478107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488107 Supplier Thermo Scientific Alfa Aesar Supplier No. H51505.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 240816-28-8
Molecular Formula C17H33NOSSn
Molecular Weight (g/mol) 418.23
MDL Number MFCD07787373
InChI Key NBICALVSSSYBKT-UHFFFAOYSA-N
Synonym 2-ethoxy-4-tributylstannyl thiazole, 2-ethoxy-4-tributylstannyl-1,3-thiazole, acmc-209g8q, tributyl-2-ethoxy-1,3-thiazol-4-yl stannane, 2-ethoxy-4-tri-n-butylstannyl thiazole
PubChem CID 2763265
SMILES CCCC[Sn](CCCC)(CCCC)C1=CSC(OCC)=N1

Specifications

Quantity 250 mg
UN Number UN2788
IUPAC Name 2-ethoxy-4-(tributylstannyl)-1,3-thiazole
Formula Weight 418.21
Percent Purity 96%
Chemical Name or Material 2-Ethoxy-4-(tri-n-butylstannyl)thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.