accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SnH3CCH3SNOH3C

CAS RN 240816-28-8

IUPAC Name: 2-ethoxy-4-(tributylstannyl)-1,3-thiazole
Synonyms: 2-ethoxy-4-(tributylstannyl)-1,3-thiazole
Molecular Weight (g/mol): 418.23
Molecular Formula: C17H33NOSSn
InChi Key: NBICALVSSSYBKT-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CSC(OCC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results