Learn More
2-Chloromethyl-3,4-dimethoxypyridine Hydrochloride, Mikromol™
Product Code. 30462370
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. 30462370
Brand: Mikromol MM0099.0500254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Chloromethyl-3,4-dimethoxypyridine Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloromethyl-3,4-dimethoxypyridine Hydrochloride
CAS 72830-09-2
Analyte or Component Names 2-Chloromethyl-3,4-dimethoxypyridine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C8 H10 Cl N O2 . Cl H
Certifications/Compliance ISO 17025
Synonym 2-Chloromethyl-3,4-dimethoxypyridine Hydrochloride
InChI Formula InChI=1S/C8H10ClNO2.ClH/c1-11-7-3-4-10-6(5-9)8(7)12-2;/h3-4H,5H2,1-2H3;1H
Solution Type Neat
SMILES Cl.COc1ccnc(CCl)c1OC
IUPAC Name 2-(chloromethyl)-3,4-dimethoxypyridine;hydrochloride
Molecular Weight (g/mol) 224.08
Quantity 4 x 25 mg
Formula Weight 223.0167 g/mol