missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloroethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific™

Product Code. p-7043386
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11370094 10 g 10g
11380094 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11370094 Supplier Thermo Scientific Alfa Aesar Supplier No. L00538.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31238-20-7
Molecular Formula C20H19BrClP
Molecular Weight (g/mol) 405.70
MDL Number MFCD00051877
InChI Key HETXOZDAMZALJQ-UHFFFAOYSA-M
Synonym 2-chloroethyl triphenylphosphonium bromide, 2-chloroethyltriphenylphosphonium bromide, 2-chloroethyl triphenylphosphanium bromide, acmc-1cp77, 2-chloroethyl triphenyl phosphanium bromide, 2-chloroethyl triphenyl phosphonium bromide, phosphonium, 2-chloroethyl triphenyl-, bromide 1:1
PubChem CID 3084530
IUPAC Name 2-chloroethyl(triphenyl)phosphanium;bromide
SMILES [Br-].ClCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 169°C to 172°C
Quantity 10 g
Sensitivity Hygroscopic
Formula Weight 405.7
Percent Purity ≥98%
Chemical Name or Material (2-Chloroethyl)triphenylphosphonium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.