accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrClP

CAS RN 31238-20-7

IUPAC Name: (2-chloroethyl)triphenylphosphanium bromide
Synonyms: (2-chloroethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 405.7
Molecular Formula: C20H19BrClP
InChi Key: HETXOZDAMZALJQ-UHFFFAOYSA-M
SMILES: [Br-].ClCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results