Learn More
2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate, Mikromol™
Product Code. 30463151
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463151 4 x 25 mg -
17943666 100 mg 100mg
17980586 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30463151 Supplier Mikromol Supplier No. MM0386.1200254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate
CAS 2727065-71-4
Analyte or Component Names 2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate
Shipping Condition Room Temperature
Molecular Formula C10 H10 Cl N3 O2 . C H4 O3 S
Certifications/Compliance ISO 17025
Synonym Doxazosin Mesilate Imp. F (EP), Doxazosin USP Related Compound C, Doxazosin USP RC C, 2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate, Doxazosin Mesilate Imp. F (EP) as Mesilate, Doxazosin Related Compound C as Mesylate, Doxazosin Mesilate Impurity F as Mesilate, Doxazosin Impurity F as Mesilate
InChI Formula InChI=1S/C11H9Cl4NO2/c12-5-8(16-11(18)10(14)15)9(17)6-1-3-7(13)4-2-6/h1-4,8,10H,5H2,(H,16,18)
Solution Type Neat
SMILES COc1cc2nc(Cl)nc(N)c2cc1OC.CS(=O)(=O)O
IUPAC Name 2-chloro-6,7-dimethoxyquinazolin-4-amine;methanesulfonic acid
Molecular Weight (g/mol) 335.76
Quantity 4 x 25 mg
Formula Weight 335.0343 g/mol