Learn More
2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate, Mikromol™
Description
2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate |
| CAS | 2727065-71-4 |
| Analyte or Component Names | 2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H10 Cl N3 O2 . C H4 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Doxazosin Mesilate Imp. F (EP), Doxazosin USP Related Compound C, Doxazosin USP RC C, 2-Chloro-6,7-dimethoxyquinazolin-4-amine Mesilate, Doxazosin Mesilate Imp. F (EP) as Mesilate, Doxazosin Related Compound C as Mesylate, Doxazosin Mesilate Impurity F as Mesilate, Doxazosin Impurity F as Mesilate |
| InChI Formula | InChI=1S/C11H9Cl4NO2/c12-5-8(16-11(18)10(14)15)9(17)6-1-3-7(13)4-2-6/h1-4,8,10H,5H2,(H,16,18) |
| SMILES | COc1cc2nc(Cl)nc(N)c2cc1OC.CS(=O)(=O)O |
| IUPAC Name | 2-chloro-6,7-dimethoxyquinazolin-4-amine;methanesulfonic acid |
| Show More |