missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-hydroxybenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 15463387
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463387 250 mg 250mg
11382089 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463387 Supplier Thermo Scientific Alfa Aesar Supplier No. H26112.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56961-30-9
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD04038818
InChI Key UTVCLUZQPSRKMY-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxybenzoicacid, benzoic acid, 2-chloro-5-hydroxy, 3-carboxy-4-chlorophenol, 2-chloro-5-hydroxybenzenecarboxylic acid, 2-chloro-5-hydroxy-benzoic acid, 2-chloro-5-hydroxy benzoic acid, 2-chloro-5-hydroxybenzene carboxylic acid, pubchem16487, acmc-1au79, ksc269a4t
PubChem CID 458210
IUPAC Name 2-chloro-5-hydroxybenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC(O)=C1

Specifications

Melting Point 178°C to 180°C
Quantity 250 mg
Formula Weight 172.57
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-hydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.