missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-iodonicotinic acid, 98%, Thermo Scientific Chemicals

Product Code. p-7033042
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414440 1 g 1g
11424440 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414440 Supplier Thermo Scientific Alfa Aesar Supplier No. H50049.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2-Chloro-4-iodonicotinic acid is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-iodonicotinic acid is used as a pharmaceutical intermediates.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544671-78-5
Molecular Formula C6H3ClINO2
Molecular Weight (g/mol) 283.45
MDL Number MFCD05663520
InChI Key XOMBFOCJXYZRSI-UHFFFAOYSA-N
Synonym 2-chloro-4-iodonicotinic acid, 2-chloro-4-iodo-nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-4-iodo, acmc-20a5ow, 2-chloro-4-iodo-3-pyridinecarboxylic acid, 3-pyridinecarboxylicacid,2-chloro-4-iodo, 2-chloro-4-iodopyridine-3-carboxylic acid
PubChem CID 17750174
IUPAC Name 2-chloro-4-iodopyridine-3-carboxylic acid
SMILES OC(=O)C1=C(Cl)N=CC=C1I

Specifications

Melting Point 162°C to 167°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 283.45
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-iodonicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.