accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOClNI

CAS RN 544671-78-5

IUPAC Name: 2-chloro-4-iodopyridine-3-carboxylic acid
Synonyms: 2-chloro-4-iodopyridine-3-carboxylic acid
Molecular Weight (g/mol): 283.45
Molecular Formula: C6H3ClINO2
InChi Key: XOMBFOCJXYZRSI-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)N=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results