Learn More
2-Chloro-3-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™
Product Code. 11346679
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346679 1 g 1g
11356679 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11346679 Supplier Thermo Scientific Alfa Aesar Supplier No. H26232.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62584-32-1
Molecular Formula C8H3ClF3N
Molecular Weight (g/mol) 205.56
MDL Number MFCD04115989
InChI Key GANPUEIPQFDYQA-UHFFFAOYSA-N
Synonym 2-chloro-3-trifluoromethyl benzonitrile, benzonitrile, 2-chloro-3-trifluoromethyl, acmc-209ws0, ksc828m3p, 2-chloro-3-cyanobenzotrifluoride, 2-chloro-3-trifluoromethyl benzenecarbonitrile
PubChem CID 3606839
IUPAC Name 2-chloro-3-(trifluoromethyl)benzonitrile
SMILES FC(F)(F)C1=CC=CC(C#N)=C1Cl
Specifications
Melting Point 38°C to 41°C
Quantity 1 g
UN Number UN3439
Formula Weight 205.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzonitrile