CAS RN 62584-32-1
CAS RN 62584-32-1
IUPAC Name:
2-chloro-3-(trifluoromethyl)benzonitrile
Synonyms:
2-chloro-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol):
205.56
Molecular Formula:
C8H3ClF3N
InChi Key:
GANPUEIPQFDYQA-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=CC(C#N)=C1Cl
1
–
1
of
1
results
1
2-Chloro-3-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™
CAS: 62584-32-1 Molecular Formula: C8H3ClF3N Molecular Weight (g/mol): 205.56 MDL Number: MFCD04115989 InChI Key: GANPUEIPQFDYQA-UHFFFAOYSA-N Synonym: 2-chloro-3-trifluoromethyl benzonitrile,benzonitrile, 2-chloro-3-trifluoromethyl,acmc-209ws0,ksc828m3p,2-chloro-3-cyanobenzotrifluoride,2-chloro-3-trifluoromethyl benzenecarbonitrile PubChem CID: 3606839 IUPAC Name: 2-chloro-3-(trifluoromethyl)benzonitrile SMILES: FC(F)(F)C1=CC=CC(C#N)=C1Cl