missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene, 98%

Product Code. p-7055602
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488776 1 g 1g
15498776 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488776 Supplier Thermo Scientific Alfa Aesar Supplier No. H64221.03
Add to basket
This item is not returnable. View return policy

CAS: 654-55-7 | C8H2ClF6NO2 | 293.549 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654-55-7
Molecular Formula C8H2ClF6NO2
Molecular Weight (g/mol) 293.549
MDL Number MFCD04972677
InChI Key OCRFDZOBCZLPRB-UHFFFAOYSA-N
Synonym 2-chloro-1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl-2-chloronitrobenzene, 3,5-bis trifluoromethyl-2-chloro-nitrobenzene, 2-chloro-3,5-di trifluoromethyl nitrobenzene, 2-chloro-3,5-bis trifluoromethyl nitrobenzene, pubchem16471, 2,4-bis-trifluoromethyl-6-nitrochlorobenzene, 2-chloro-3,5-bis-trifluoromethyl-nitrobenzene, 3,5-bis-trifluoromethyl-2-chloro-nitrobenzene, 2-chloro-1-nitro-3,5-bis-trifluoromethyl-benzene
PubChem CID 10803688
IUPAC Name 2-chloro-1-nitro-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1C(F)(F)F)Cl)[N+](=O)[O-])C(F)(F)F

Specifications

Quantity 1 g
Beilstein 2888862
Formula Weight 293.55
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.