accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 654-55-7

FFFClNOOFFF

CAS RN 654-55-7

IUPAC Name: 2-chloro-1-nitro-3,5-bis(trifluoromethyl)benzene
Synonyms: 2-chloro-1-nitro-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 293.55
Molecular Formula: C8H2ClF6NO2
InChi Key: OCRFDZOBCZLPRB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC(=C1Cl)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results