missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,4-di-n-propoxybenzene, 97%, Thermo Scientific™

Product Code. 11476506
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476506 10 g 10g
11486506 50 g 50g
11496506 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476506 Supplier Thermo Scientific Alfa Aesar Supplier No. B24776.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916791-98-5
Molecular Formula C12H17ClO2
Molecular Weight (g/mol) 228.716
MDL Number MFCD00048688
InChI Key SWQFWWUXVCCQSJ-UHFFFAOYSA-N
Synonym 2-chloro-1,4-di-n-propoxybenzene, chlorohydroquinone di-n-propyl ether, 1-chloro-2,5-di-n-propoxybenzene
PubChem CID 7020403
IUPAC Name 2-chloro-1,4-dipropoxybenzene
SMILES CCCOC1=CC(=C(C=C1)OCCC)Cl

Specifications

Quantity 10 g
Refractive Index 1.513
Formula Weight 228.72
Percent Purity 97%
Chemical Name or Material 2-Chloro-1,4-di-n-propoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.