accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 916791-98-5

IUPAC Name: 2-chloro-1,4-dipropoxybenzene
Synonyms: 2-chloro-1,4-dipropoxybenzene
Molecular Weight (g/mol): 228.72
Molecular Formula: C12H17ClO2
InChi Key: SWQFWWUXVCCQSJ-UHFFFAOYSA-N
SMILES: CCCOC1=CC(Cl)=C(OCCC)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results